COMH4211BB18 Combi-Blocks, Inc.

1-Phenyl-2,5,8,11-tetraoxatridecan-13-yl 4-methylbenzenesulfonate

  • MDL number MFCD29060061
  • Cas Number : 89346-82-7
  • Molecular Weight : 438.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6482

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6482
Availability details for
Quantity
  • Availability
  • Add to Cart