COMH4211BB19 Combi-Blocks, Inc.

2-Chloro-4-(3-methyl-1h-1,2,4-triazol-1-yl)benzaldehyde

  • MDL number MFCD31579446
  • Cas Number : 1819358-30-9
  • Molecular Weight : 221.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6484

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6484
Availability details for
Quantity
  • Availability
  • Add to Cart