COMH4211BB1A Combi-Blocks, Inc.

1-(6-Aminopyridin-3-yl)-3-methylazetidin-3-ol

  • MDL number MFCD24644910
  • Cas Number : 1253911-28-2
  • Molecular Weight : 179.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6485

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6485
Availability details for
Quantity
  • Availability
  • Add to Cart