COMH4211BB1D Combi-Blocks, Inc.

(6-Cyclopropanesulfonyl-pyridin-3-yl)-methanol

  • MDL number MFCD27937145
  • Cas Number : 1823896-34-9
  • Molecular Weight : 213.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (6-(cyclopropylsulfonyl)pyridin-3-yl)methanol
Inchi Code 1S/C9H11NO3S/c11-6-7-1-4-9(10-5-7)14(12,13)8-2-3-8/h1,4-5,8,11H,2-3,6H2
Inchi Key BSRFMQNXNFKZTM-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6491

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6491
Availability details for
Quantity
  • Availability
  • Add to Cart