COMH4211BB20 Combi-Blocks, Inc.

1-(4-Bromo-2-cyclopropylmethoxy-phenyl)-ethylamine

  • MDL number MFCD24341520
  • Cas Number : 1552615-62-9
  • Molecular Weight : 270.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6501

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6501
Availability details for
Quantity
  • Availability
  • Add to Cart