COMH4211BB22 Combi-Blocks, Inc.

1-Bromo-4-(1-bromo-2-methylpropyl)benzene

  • MDL number MFCD18354292
  • Cas Number : 1341871-86-0
  • Molecular Weight : 292.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6505

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6505
Availability details for
Quantity
  • Availability
  • Add to Cart