COMH4211BB23 Combi-Blocks, Inc.

4-(Pyridin-4-ylsulfanyl)aniline

  • MDL number MFCD08447091
  • Cas Number : 673-09-6
  • Molecular Weight : 202.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6507

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6507
Availability details for
Quantity
  • Availability
  • Add to Cart