COMH4211BB24 Combi-Blocks, Inc.

Toluene-4-sulfonic acid 2-(2-2-[2-(tert-butyl-diphenyl-silanyloxy)-ethoxy]-ethoxy-ethoxy)-ethyl ester

  • MDL number MFCD31614128
  • Cas Number : 859385-34-5
  • Molecular Weight : 586.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6513

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6513
Availability details for
Quantity
  • Availability
  • Add to Cart