COMH4211BB26 Combi-Blocks, Inc.

3-Fluoro-4-(2-hydroxyethoxy)benzonitrile

  • MDL number MFCD12110271
  • Cas Number : 917226-74-5
  • Molecular Weight : 181.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-fluoro-4-(2-hydroxyethoxy)benzonitrile
Inchi Code 1S/C9H8FNO2/c10-8-5-7(6-11)1-2-9(8)13-4-3-12/h1-2,5,12H,3-4H2
Inchi Key XSUCKXYFTBYZMV-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6518

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6518
Availability details for
Quantity
  • Availability
  • Add to Cart