COMH4211BB27 Combi-Blocks, Inc.

2-(3-Iodo-phenyl)-isothiazolidine 1,1-dioxide

  • MDL number MFCD14659212
  • Cas Number : 1228776-05-3
  • Molecular Weight : 323.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6521

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6521
Availability details for
Quantity
  • Availability
  • Add to Cart