COMH4211BB29 Combi-Blocks, Inc.

(R)-(+)-5-(2-Aminopropyl)-2-methoxybenzene sulfonamide hydrochloride

  • MDL number MFCD08460105
  • Cas Number : 112101-75-4
  • Molecular Weight : 280.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6525

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6525
Availability details for
Quantity
  • Availability
  • Add to Cart