COMH4211BB2B Combi-Blocks, Inc.

4-((6-Bromopyridin-3-yl)oxy)benzonitrile

  • MDL number MFCD22987899
  • Cas Number : 1207731-13-2
  • Molecular Weight : 275.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6528

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6528
Availability details for
Quantity
  • Availability
  • Add to Cart