COMH4211BB2F Combi-Blocks, Inc.

3-Amino-5-fluoro-n-(prop-2-en-1-yl)benzamide

  • MDL number MFCD24125294
  • Cas Number : 1515465-66-3
  • Molecular Weight : 194.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6535

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6535
Availability details for
Quantity
  • Availability
  • Add to Cart