COMH4211BB30 Combi-Blocks, Inc.

2-Bromo-4-iodo-benzoic acid benzyl ester

  • MDL number MFCD30742726
  • Cas Number : 2206265-51-0
  • Molecular Weight : 417.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name benzyl 2-bromo-4-iodobenzoate
Inchi Code 1S/C14H10BrIO2/c15-13-8-11(16)6-7-12(13)14(17)18-9-10-4-2-1-3-5-10/h1-8H,9H2
Inchi Key HROMYJZLBAQDKD-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6537

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6537
Availability details for
Quantity
  • Availability
  • Add to Cart