COMH4211BB31 Combi-Blocks, Inc.

5-Amino-n,2-dimethylbenzamide

  • MDL number MFCD14618173
  • Cas Number : 1249563-99-2
  • Molecular Weight : 164.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6542

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6542
Availability details for
Quantity
  • Availability
  • Add to Cart