COMH4211BB33 Combi-Blocks, Inc.

4-Chloro-2-[4-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-pyrazol-1-ylmethyl]-benzoic acid methyl ester

  • MDL number MFCD31583332
  • Molecular Weight : 376.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 4-chloro-2-((4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)methyl)benzoate
Inchi Code 1S/C18H22BClN2O4/c1-17(2)18(3,4)26-19(25-17)13-9-21-22(11-13)10-12-8-14(20)6-7-15(12)16(23)24-5/h6-9,11H,10H2,1-5H3
Inchi Key FKSWDLXGSYPANP-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6545

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6545
Availability details for
Quantity
  • Availability
  • Add to Cart