COMH4211BB34 Combi-Blocks, Inc.

3,5-Dichloro-4-(3-fluoro-propoxy)-benzaldehyde

  • MDL number MFCD31628768
  • Cas Number : 1562412-70-7
  • Molecular Weight : 251.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6547

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6547
Availability details for
Quantity
  • Availability
  • Add to Cart