COMH4211BB35 Combi-Blocks, Inc.

Benzeneacetamide, 4-methyl-n-[4-(5-oxazolyl)phenyl]-

  • MDL number MFCD30486225
  • Cas Number : 1626182-01-1
  • Molecular Weight : 292.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6548

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6548
Availability details for
Quantity
  • Availability
  • Add to Cart