COMH4211BB37 Combi-Blocks, Inc.

4-[(Propan-2-ylsulfanyl)methyl]piperidine hydrochloride

  • MDL number MFCD26793160
  • Cas Number : 1366664-45-0
  • Molecular Weight : 209.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6551

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6551
Availability details for
Quantity
  • Availability
  • Add to Cart