COMH4211BB39 Combi-Blocks, Inc.

4-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-1-(3-trifluoromethyl-benzyl)-1h-pyrazole

  • MDL number MFCD21337504
  • Cas Number : 1415825-08-9
  • Molecular Weight : 352.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6560

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6560
Availability details for
Quantity
  • Availability
  • Add to Cart