COMH4211BB3C Combi-Blocks, Inc.

2-Amino-n,n-dimethyl-butyramide oxalate

  • MDL number MFCD29763675
  • Cas Number : 2365418-06-8
  • Molecular Weight : 220.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6569

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6569
Availability details for
Quantity
  • Availability
  • Add to Cart