COMH4211BB3D Combi-Blocks, Inc.

(3-Bromo-5-trifluoromethyl-phenyl)-pyrrolidin-1-yl-methanone

  • MDL number MFCD31619010
  • Molecular Weight : 322.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6572

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6572
Availability details for
Quantity
  • Availability
  • Add to Cart