COMH4211BB40 Combi-Blocks, Inc.

4-(1H-Pyrazol-1-yl)butan-1-ol

  • MDL number MFCD12186770
  • Cas Number : 1156067-60-5
  • Molecular Weight : 140.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6579

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6579
Availability details for
Quantity
  • Availability
  • Add to Cart