COMH4211BB42 Combi-Blocks, Inc.

5-Bromo-2-cyclobutoxy-4-methyl-pyrimidine

  • MDL number MFCD31583178
  • Cas Number : 1935588-15-0
  • Molecular Weight : 243.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6582

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6582
Availability details for
Quantity
  • Availability
  • Add to Cart