COMH4211BB44 Combi-Blocks, Inc.

[2-Methyl-4-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-phenyl]-pyrrolidin-1-yl-methanone

  • MDL number MFCD26399448
  • Cas Number : 1092563-41-1
  • Molecular Weight : 315.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)(pyrrolidin-1-yl)methanone
Inchi Code 1S/C18H26BNO3/c1-13-12-14(19-22-17(2,3)18(4,5)23-19)8-9-15(13)16(21)20-10-6-7-11-20/h8-9,12H,6-7,10-11H2,1-5H3
Inchi Key SWJZMZPRCHGWGV-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6584

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6584
Availability details for
Quantity
  • Availability
  • Add to Cart