COMH4211BB45 Combi-Blocks, Inc.

1-((2-(Trimethylsilyl)ethoxy)methyl)-1h-pyrazol-4-ol

  • MDL number MFCD31579310
  • Cas Number : 220299-50-3
  • Molecular Weight : 214.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6585

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6585
Availability details for
Quantity
  • Availability
  • Add to Cart