COMH4211BB48 Combi-Blocks, Inc.

(R)-3-Methyl-2-(3-methylbenzamido)butanoic acid

  • MDL number MFCD21541596
  • Cas Number : 1608722-14-0
  • Molecular Weight : 235.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6596

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6596
Availability details for
Quantity
  • Availability
  • Add to Cart