COMH4211BB4B Combi-Blocks, Inc.

4-Iodo-n-(2,2,2-trifluoroethyl)benzamide

  • MDL number MFCD14644365
  • Cas Number : 1249062-31-4
  • Molecular Weight : 329.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6607

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6607
Availability details for
Quantity
  • Availability
  • Add to Cart