COMH4211BB4D Combi-Blocks, Inc.

6-Amino-n-(2,2,2-trifluoro-ethyl)-nicotinamide

  • MDL number MFCD16678975
  • Cas Number : 1249487-67-9
  • Molecular Weight : 219.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6611

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6611
Availability details for
Quantity
  • Availability
  • Add to Cart