COMH4211BB4E Combi-Blocks, Inc.

2-(2-Chloroethoxy)benzaldehyde

  • MDL number MFCD01961271
  • Cas Number : 54373-14-7
  • Molecular Weight : 184.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6612

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6612
Availability details for
Quantity
  • Availability
  • Add to Cart