COMH4211BB50 Combi-Blocks, Inc.

3,5-Bis-(tert-butyl-dimethyl-silanyloxy)-phenol

  • MDL number MFCD31614134
  • Cas Number : 197248-10-5
  • Molecular Weight : 354.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6614

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6614
Availability details for
Quantity
  • Availability
  • Add to Cart