COMH4211BB52 Combi-Blocks, Inc.

1-(3-Nitro-4-propoxyphenyl)ethan-1-one

  • MDL number MFCD11619315
  • Cas Number : 54375-49-4
  • Molecular Weight : 223.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6616

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6616
Availability details for
Quantity
  • Availability
  • Add to Cart