COMH4211BB54 Combi-Blocks, Inc.

2-(3-Iodophenoxy)-n,n-dimethylethanamine

  • MDL number MFCD14648745
  • Cas Number : 103808-65-7
  • Molecular Weight : 291.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6620

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6620
Availability details for
Quantity
  • Availability
  • Add to Cart