COMH4211BB56 Combi-Blocks, Inc.

5-Bromo-2-cyclopropylmethoxy-4-methyl-pyridine

  • MDL number MFCD31613894
  • Cas Number : 1600268-01-6
  • Molecular Weight : 242.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6625

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6625
Availability details for
Quantity
  • Availability
  • Add to Cart