COMH4211BB59 Combi-Blocks, Inc.

4-Fluoro-3-(trifluoromethyl)phenylurea

  • MDL number MFCD14600405
  • Cas Number : 1000588-77-1
  • Molecular Weight : 222.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-[4-fluoro-3-(trifluoromethyl)phenyl]urea
Inchi Code 1S/C8H6F4N2O/c9-6-2-1-4(14-7(13)15)3-5(6)8(10,11)12/h1-3H,(H3,13,14,15)
Inchi Key QLDISJBNQHJOHZ-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6639

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6639
Availability details for
Quantity
  • Availability
  • Add to Cart