COMH4211BB5B Combi-Blocks, Inc.

3-(4-Piperidinylmethyl)-benzoic acid methyl ester

  • MDL number MFCD22403353
  • Cas Number : 1170775-45-7
  • Molecular Weight : 233.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6641

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6641
Availability details for
Quantity
  • Availability
  • Add to Cart