COMH4211BB5C Combi-Blocks, Inc.

6-Isobutoxy-1h-indazol-3-amine

  • MDL number MFCD28975974
  • Cas Number : 1394068-70-2
  • Molecular Weight : 205.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6643

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6643
Availability details for
Quantity
  • Availability
  • Add to Cart