COMH4211BB5D Combi-Blocks, Inc.

[2-Bromo-3-(propan-2-yloxy)phenyl]methanamine

  • MDL number MFCD31613856
  • Cas Number : 1936662-46-2
  • Molecular Weight : 244.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6646

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6646
Availability details for
Quantity
  • Availability
  • Add to Cart