COMH4211BB60 Combi-Blocks, Inc.

(R)-2-Amino-1-(pyrrolidin-1-yl)butan-1-one

  • MDL number MFCD21296589
  • Cas Number : 1258066-46-4
  • Molecular Weight : 156.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2R)-1-oxo-1-(1-pyrrolidinyl)-2-butanamine
Inchi Code 1S/C8H16N2O/c1-2-7(9)8(11)10-5-3-4-6-10/h7H,2-6,9H2,1H3/t7-/m1/s1
Inchi Key LSMYTQPMEHMSCJ-SSDOTTSWSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6654

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6654
Availability details for
Quantity
  • Availability
  • Add to Cart