COMH4211BB61 Combi-Blocks, Inc.

3-(4-Acetyl-2-trifluoromethyl-phenoxy)-piperidine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD31539576
  • Cas Number : 1100768-25-9
  • Molecular Weight : 387.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6655

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6655
Availability details for
Quantity
  • Availability
  • Add to Cart