COMH4211BB62 Combi-Blocks, Inc.

N-Methyl moxifloxacin hcl

  • MDL number MFCD23704648
  • Cas Number : 721970-37-2
  • Molecular Weight : 451.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6656

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6656
Availability details for
Quantity
  • Availability
  • Add to Cart