COMH4211BB63 Combi-Blocks, Inc.

3-(Benzyloxy)pyridin-2-ol

  • MDL number MFCD26792765
  • Cas Number : 94475-64-6
  • Molecular Weight : 201.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(benzyloxy)-2(1H)-pyridinone
Inchi Code 1S/C12H11NO2/c14-12-11(7-4-8-13-12)15-9-10-5-2-1-3-6-10/h1-8H,9H2,(H,13,14)
Inchi Key CHPFCHGKXZKMHV-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6657

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6657
Availability details for
Quantity
  • Availability
  • Add to Cart