COMH4211BB67 Combi-Blocks, Inc.

(R)-2-(2-Fluoro-5-(trifluoromethyl)benzamido)-3-methylbutanoic acid

  • MDL number MFCD31579570
  • Molecular Weight : 307.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6666

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6666
Availability details for
Quantity
  • Availability
  • Add to Cart