COMH4211BB6A Combi-Blocks, Inc.

3-(1-Methanesulfonyl-piperidin-4-yl)-propionic acid

  • MDL number MFCD16692943
  • Cas Number : 1250825-72-9
  • Molecular Weight : 235.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-[1-(methylsulfonyl)-4-piperidinyl]propanoic acid
Inchi Code 1S/C9H17NO4S/c1-15(13,14)10-6-4-8(5-7-10)2-3-9(11)12/h8H,2-7H2,1H3,(H,11,12)
Inchi Key CJFRODIZZIJRGE-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6670

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6670
Availability details for
Quantity
  • Availability
  • Add to Cart