COMH4211BB6B Combi-Blocks, Inc.

4-(4-tert-Butoxycarbonylamino-phenyl)-2,2-dimethyl-butyric acid

  • MDL number MFCD31613962
  • Cas Number : 1398047-46-5
  • Molecular Weight : 307.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6671

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6671
Availability details for
Quantity
  • Availability
  • Add to Cart