COMH4211BB70 Combi-Blocks, Inc.

2-(Cyclopropylmethoxy)-5-hydroxy-n-methylbenzamide

  • MDL number MFCD16998410
  • Cas Number : 1243475-82-2
  • Molecular Weight : 221.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6690

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6690
Availability details for
Quantity
  • Availability
  • Add to Cart