COMH4211BB71 Combi-Blocks, Inc.

O-Cyclohexylmethyl-n-methyl-hydroxylamine hydrochloride

  • MDL number MFCD31561572
  • Cas Number : 2270906-60-8
  • Molecular Weight : 179.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6691

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6691
Availability details for
Quantity
  • Availability
  • Add to Cart