COMH4211BB73 Combi-Blocks, Inc.

Benzoic acid, 4-[[(phenylmethoxy)amino]methyl]-, methyl ester

  • MDL number MFCD31561570
  • Cas Number : 1493447-94-1
  • Molecular Weight : 271.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6694

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6694
Availability details for
Quantity
  • Availability
  • Add to Cart