COMH4211BB74 Combi-Blocks, Inc.

(2R)-2-Methyl-1-(propane-2-sulfonyl)piperazine

  • MDL number MFCD24301085
  • Cas Number : 1568031-81-1
  • Molecular Weight : 206.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6695

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6695
Availability details for
Quantity
  • Availability
  • Add to Cart