COMH4211BB76 Combi-Blocks, Inc.

3-(2-Methyl-2,3-dihydro-1h-indol-1-yl)propan-1-amine

  • MDL number MFCD09815186
  • Cas Number : 1016529-13-7
  • Molecular Weight : 190.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6700

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6700
Availability details for
Quantity
  • Availability
  • Add to Cart